siRNA Details
| Gene Symbol: Tmem98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007672.1 | NP_001007673.1 | 7 | 1378 | ||
| GenBank mRNA | BC078932.1 | - | 7 | 1378 | 7107480 | |
| FQ214419.1 | - | 7 | 1533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem98| Gene Symbol: Tmem98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007672.1 | NP_001007673.1 | 7 | 1378 | ||
| GenBank mRNA | BC078932.1 | - | 7 | 1378 | 7107480 | |
| FQ214419.1 | - | 7 | 1533 | |||