siRNA Details
| Gene Symbol: Slc25a19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007674.1 | NP_001007675.1 | 7 | 1316 | ||
| GenBank mRNA | BC079002.1 | - | 7 | 1316 | 7112815 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a19| Gene Symbol: Slc25a19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007674.1 | NP_001007675.1 | 7 | 1316 | ||
| GenBank mRNA | BC079002.1 | - | 7 | 1316 | 7112815 | |