siRNA Details
| Gene Symbol: C1qtnf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007675.1 | NP_001007676.1 | 4 | 1059 | ||
| XM_006247800.2 | XP_006247862.1 | 4 | 1585 | |||
| XM_006247801.2 | XP_006247863.1 | 4 | 1200 | |||
| XM_006247802.2 | XP_006247864.1 | 4 | 1657 | |||
| XM_008768374.1 | XP_008766596.1 | 4 | 1261 | |||
| GenBank mRNA | BC083720.1 | - | 4 | 1059 | 7190841 | |
| FQ209955.1 | - | 4 | 1160 | |||

C1qtnf1
Genomic Map