siRNA Details
| Gene Symbol: Zfp219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007681.1 | NP_001007682.1 | 4 | 2609 | ||
| XM_006251878.2 | XP_006251940.1 | 5 | 3768 | |||
| XM_006251879.2 | XP_006251941.1 | 6 | 3426 | |||
| XM_006251880.2 | XP_006251942.1 | 5 | 3152 | |||
| XM_006251882.2 | XP_006251944.1 | 4 | 3063 | |||
| XM_008770522.1 | XP_008768744.1 | 5 | 3452 | |||
| GenBank mRNA | BC082017.1 | - | 4 | 2609 | 7132141 | |
| FQ212172.1 | - | 5 | 2592 | |||
| FQ221971.1 | - | 6 | 3016 | |||

Zfp219
Genomic Map