siRNA Details
| Gene Symbol: Zfp219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007681.1 | NP_001007682.1 | 2 | 1122 | ||
| XM_006251878.2 | XP_006251940.1 | 3 | 2281 | |||
| XM_006251879.2 | XP_006251941.1 | 4 | 1939 | |||
| XM_006251880.2 | XP_006251942.1 | 3 | 1665 | |||
| XM_006251882.2 | XP_006251944.1 | 2 | 1576 | |||
| XM_008770522.1 | XP_008768744.1 | 3 | 1965 | |||
| GenBank mRNA | BC082017.1 | - | 2 | 1122 | 7132141 | |
| FQ212172.1 | - | 3 | 1105 | |||
| FQ221971.1 | - | 4 | 1529 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001253695, NM_001253696, NM_001253694, NM_027248, BC052050, and others.

Zfp219
Genomic Map