siRNA Details
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007682.1 | NP_001007683.1 | 2 | 1140 | ||
GenBank mRNA | BC081978.1 | - | 2 | 1140 | 7128058 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Thtpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007682.1 | NP_001007683.1 | 2 | 1140 | ||
GenBank mRNA | BC081978.1 | - | 2 | 1140 | 7128058 |