siRNA Details
| Gene Symbol: Klf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007684.1 | NP_001007685.1 | 2 | 841 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klf2| Gene Symbol: Klf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007684.1 | NP_001007685.1 | 2 | 841 | ||