siRNA Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007685.1 | NP_001007686.1 | 6 | 1294 | ||
| XM_008766516.1 | XP_008764738.1 | 6 | 1688 | |||
| XM_008766517.1 | XP_008764739.1 | 6 | 1381 | |||
| GenBank mRNA | BC083777.1 | - | 6 | 1294 | 7193594 | |

Klhdc8b
Genomic Map