siRNA Details
| Gene Symbol: Cln8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007686.1 | NP_001007687.1 | 3 | 1092 | ||
| XM_006253394.2 | XP_006253456.1 | 3 | 1366 | |||
| GenBank mRNA | BC078982.1 | - | 3 | 1092 | 7112147 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cln8| Gene Symbol: Cln8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007686.1 | NP_001007687.1 | 3 | 1092 | ||
| XM_006253394.2 | XP_006253456.1 | 3 | 1366 | |||
| GenBank mRNA | BC078982.1 | - | 3 | 1092 | 7112147 | |