siRNA Details
| Gene Symbol: Ciapin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007689.1 | NP_001007690.1 | 5 | 801 | ||
| XM_006255106.2 | XP_006255168.1 | 5 | 888 | |||
| XM_006255107.2 | XP_006255169.1 | 5 | 600 | |||
| XM_006255108.2 | XP_006255170.1 | 5 | 603 | |||
| GenBank mRNA | BC083753.1 | - | 5 | 801 | 7192604 | |
| FQ211546.1 | - | 5 | 585 | |||
| FQ213033.1 | - | 5 | 632 | |||
| FQ230885.1 | - | 5 | 813 | |||

Ciapin1
Genomic Map