siRNA Details
| Gene Symbol: M6pr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007700.1 | NP_001007701.1 | 3 | 467 | ||
| XM_008763247.1 | XP_008761469.1 | 3 | 1519 | |||
| XM_008763248.1 | XP_008761470.1 | 3 | 532 | |||
| GenBank mRNA | BC079226.1 | - | 3 | 467 | 7132318 | |
| Gene Symbol: LOC100909548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752100.1 | XP_003752148.1 | 3 | 473 | ||
| XM_006257201.2 | XP_006257263.1 | 3 | 1519 | |||
| XM_006257202.1 | XP_006257264.1 | 3 | 692 | |||

M6pr
Genomic Map