siRNA Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007704.1 | NP_001007705.1 | 7 | 1257 | ||
| GenBank mRNA | BC081876.1 | - | 7 | 1257 | 7107751 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galm| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007704.1 | NP_001007705.1 | 7 | 1257 | ||
| GenBank mRNA | BC081876.1 | - | 7 | 1257 | 7107751 | |