siRNA Details
| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007711.1 | NP_001007712.1 | 4 | 336 | ||
| XM_006244321.2 | XP_006244383.1 | 5 | 459 | |||
| GenBank mRNA | BC079136.1 | - | 4 | 336 | 7127699 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf2f1| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007711.1 | NP_001007712.1 | 4 | 336 | ||
| XM_006244321.2 | XP_006244383.1 | 5 | 459 | |||
| GenBank mRNA | BC079136.1 | - | 4 | 336 | 7127699 | |