siRNA Details
| Gene Symbol: Tmbim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007713.1 | NP_001007714.1 | 11 | 1637 | ||
| XM_008767238.1 | XP_008765460.1 | 11 | 4009 | |||
| XM_008767239.1 | XP_008765461.1 | 12 | 1660 | |||
| GenBank mRNA | BC079087.1 | - | 11 | 1637 | 7123233 | |

Tmbim1
Genomic Map