siRNA Details
| Gene Symbol: Icam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007725.1 | NP_001007726.1 | 6 | 872 | ||
| XM_006247624.2 | XP_006247686.1 | 6 | 882 | |||
| XM_006247627.2 | XP_006247689.1 | 4 | 716 | |||
| XM_008768377.1 | XP_008766599.1 | 5 | 1479 | |||
| XM_008768378.1 | XP_008766600.1 | 7 | 931 | |||
| XM_008768379.1 | XP_008766601.1 | 5 | 791 | |||
| GenBank mRNA | BC079465.1 | - | 6 | 872 | 7122370 | |
| FQ220624.1 | - | 3 | 578 | |||
| FQ231566.1 | - | 6 | 741 | |||
| FQ231887.1 | - | 6 | 740 | |||

Icam2
Genomic Map