siRNA Details
| Gene Symbol: Serpinb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007732.1 | NP_001007733.1 | 7 | 910 | ||
| XM_006253872.2 | XP_006253934.1 | 7 | 879 | |||
| GenBank mRNA | BC079063.1 | - | 7 | 910 | 7121006 | |
| FQ226618.1 | - | 4 | 445 | |||

Serpinb9
Genomic Map