siRNA Details
| Gene Symbol: Sertad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007735.1 | NP_001007736.1 | 2 | 1022 | ||
| GenBank mRNA | BC061808.1 | - | 2 | 1022 | 5598343 | |
| FQ214111.1 | - | 2 | 1036 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sertad1| Gene Symbol: Sertad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007735.1 | NP_001007736.1 | 2 | 1022 | ||
| GenBank mRNA | BC061808.1 | - | 2 | 1022 | 5598343 | |
| FQ214111.1 | - | 2 | 1036 | |||