siRNA Details
| Gene Symbol: Tmem109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007736.1 | NP_001007737.1 | 2 | 297 | ||
| XM_006231059.2 | XP_006231121.1 | 3 | 568 | |||
| XM_008760235.1 | XP_008758457.1 | 3 | 475 | |||
| XM_008760236.1 | XP_008758458.1 | 2 | 475 | |||
| XM_008760237.1 | XP_008758459.1 | 3 | 692 | |||
| GenBank mRNA | BC078955.1 | - | 2 | 297 | 7110380 | |
| FQ225946.1 | - | 3 | 463 | |||

Tmem109
Genomic Map