siRNA Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007742.1 | NP_001007743.1 | 7 | 882 | ||
| XM_006233689.1 | XP_006233751.1 | 7 | 909 | |||
| GenBank mRNA | BC083603.1 | - | 7 | 882 | 7133164 | |
| FQ210767.1 | - | 7 | 768 | |||
| FQ216687.1 | - | 2 | 117 | |||

Ubac1
Genomic Map