siRNA Details
| Gene Symbol: Armcx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007757.1 | NP_001007758.1 | 3 | 519 | ||
| XM_006257228.2 | XP_006257290.1 | 4 | 640 | |||
| XM_008773423.1 | XP_008771645.1 | 7 | 1935 | |||
| XM_008773424.1 | XP_008771646.1 | 7 | 2937 | |||
| XM_008773425.1 | XP_008771647.1 | 4 | 664 | |||
| GenBank mRNA | BC081893.1 | - | 3 | 519 | 7109901 | |

Armcx6
Genomic Map