siRNA Details
| Gene Symbol: TTC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310135.1 | NP_001297064.1 | 33 | 6514 | ||
| XM_011537430.2 | XP_011535732.1 | 29 | 4346 | |||
| XM_011537431.2 | XP_011535733.1 | 27 | 4003 | |||
| XM_017021254.1 | XP_016876743.1 | 32 | 6364 | |||
| XM_017021255.1 | XP_016876744.1 | 31 | 6229 | |||
| XM_017021256.1 | XP_016876745.1 | 32 | 6628 | |||
| GenBank mRNA | BC014342.2 | - | 6 | 921 | ||
| BC103914.2 | - | 15 | 2084 | 40006180 | ||
| BC103915.1 | - | 14 | 1863 | 40006182 | ||
| BC103916.2 | - | 14 | 1863 | 40006183 | ||
| BX161415.1 | - | 13 | 1798 | |||

TTC6
Genomic Map