siRNA Details
| Gene Symbol: Glyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007800.1 | NP_001007801.1 | 3 | 114 | ||
| XM_006245803.2 | XP_006245865.1 | 3 | 183 | |||
| XM_006245804.2 | XP_006245866.1 | 3 | 183 | |||
| GenBank mRNA | BC085931.1 | - | 3 | 125 | 7111787 | |

Glyr1
Genomic Map