siRNA Details
| Gene Symbol: TATDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014760.3 | NP_055575.3 | 2 | 887 | ||
| GenBank mRNA | AK289794.1 | - | 2 | 608 | ||
| BC090935.1 | - | 2 | 468 | 3064714 | ||
| BC101770.1 | - | 2 | 376 | 8069276 | ||
| BC101776.1 | - | 2 | 377 | 8069282 | ||
| D86972.1 | - | 3 | 704 | |||

TATDN2
Genomic Map