siRNA Details
| Gene Symbol: Cmtm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007802.3 | NP_001007803.1 | 5 | 482 | ||
| XM_006243969.2 | XP_006244031.1 | 4 | 504 | |||
| GenBank mRNA | AY325262.1 | - | 5 | 482 | ||
| AY387088.1 | - | 4 | 442 | |||
| BC107930.1 | - | 4 | 475 | 7130158 | ||

Cmtm6
Genomic Map