siRNA Details
| Gene Symbol: Rtp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008230.3 | NP_001008231.1 | 2 | 795 | ||
| GenBank mRNA | BC049158.1 | - | 2 | 795 | 6390582 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtp2| Gene Symbol: Rtp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008230.3 | NP_001008231.1 | 2 | 795 | ||
| GenBank mRNA | BC049158.1 | - | 2 | 795 | 6390582 | |