siRNA Details
| Gene Symbol: TTC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008237.2 | NP_001008238.1 | 2 | 352 | ||
| GenBank mRNA | BC057850.1 | - | 2 | 352 | 4795916 | |
| BC070381.1 | - | 2 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC32| Gene Symbol: TTC32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008237.2 | NP_001008238.1 | 2 | 352 | ||
| GenBank mRNA | BC057850.1 | - | 2 | 352 | 4795916 | |
| BC070381.1 | - | 2 | 223 | |||