siRNA Details
| Gene Symbol: Lrrc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008280.1 | NP_001008281.1 | 7 | 1437 | ||
| GenBank mRNA | BC086530.1 | - | 7 | 1437 | 7304726 | |
| D13623.1 | - | 7 | 1474 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc59| Gene Symbol: Lrrc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008280.1 | NP_001008281.1 | 7 | 1437 | ||
| GenBank mRNA | BC086530.1 | - | 7 | 1437 | 7304726 | |
| D13623.1 | - | 7 | 1474 | |||