siRNA Details
| Gene Symbol: Crkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008284.1 | NP_001008285.1 | 1267 | |||
| GenBank mRNA | BC085865.1 | - | 1267 | 7190712 | ||
| Gene Symbol: LOC100911248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751095.3 | XP_003751143.1 | 2, 3 | 1299 | ||

Crkl
Genomic Map