siRNA Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008285.1 | NP_001008286.1 | 5, 6 | 422 | ||
| XM_006248331.2 | XP_006248393.1 | 6, 7 | 636 | |||
| XM_006248332.2 | XP_006248394.1 | 5, 6 | 590 | |||
| XM_006248334.1 | XP_006248396.1 | 3, 4 | 319 | |||
| XM_008768719.1 | XP_008766941.1 | 5, 6 | 520 | |||
| XM_008768721.1 | XP_008766943.1 | 3, 4 | 851 | |||
| GenBank mRNA | BC085954.1 | - | 5, 6 | 422 | 7134530 | |
| FQ210155.1 | - | 5, 6 | 488 | |||

Spice1
Genomic Map