siRNA Details
| Gene Symbol: SCRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145513.1 | NP_001138985.1 | 2 | 296 | ||
| NM_001145514.1 | NP_001138986.1 | 2 | 228 | |||
| NM_014766.4 | NP_055581.3 | 2 | 288 | |||
| GenBank mRNA | AB071705.1 | - | 2 | 178 | ||
| AK289514.1 | - | 1 | 323 | |||
| AK293462.1 | - | 2 | 240 | |||
| AK295172.1 | - | 2 | 228 | |||
| BC040492.1 | - | 2 | 240 | 5314711 | ||
| D83777.2 | - | 2 | 233 | |||
| DC307846.1 | - | 2 | 297 | |||
| DC370097.1 | - | 2 | 288 | |||

SCRN1
Genomic Map