siRNA Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008290.1 | NP_001008291.1 | 4, 5 | 649 | ||
| GenBank mRNA | BC086384.1 | - | 4, 5 | 649 | 7301624 | |
| FQ220218.1 | - | 4, 5 | 460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam216a| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008290.1 | NP_001008291.1 | 4, 5 | 649 | ||
| GenBank mRNA | BC086384.1 | - | 4, 5 | 649 | 7301624 | |
| FQ220218.1 | - | 4, 5 | 460 | |||