siRNA Details
Gene Symbol: Fam216a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008290.1 | NP_001008291.1 | 5 | 769 | ||
GenBank mRNA | BC086384.1 | - | 5 | 769 | 7301624 | |
FQ220218.1 | - | 5 | 580 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam216a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008290.1 | NP_001008291.1 | 5 | 769 | ||
GenBank mRNA | BC086384.1 | - | 5 | 769 | 7301624 | |
FQ220218.1 | - | 5 | 580 |