siRNA Details
| Gene Symbol: Diablo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008292.1 | NP_001008293.1 | 3, 4 | 338 | ||
| GenBank mRNA | BC086366.1 | - | 3, 4 | 338 | 7305418 | |
| FQ219826.1 | - | 3, 4 | 331 | |||
| Gene Symbol: LOC100360940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729540.3 | XP_002729586.2 | 2 | 368 | ||
| XM_008764180.1 | XP_008762402.1 | 1 | 155 | |||

Diablo
Genomic Map