siRNA Details
| Gene Symbol: Tfb2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008293.1 | NP_001008294.1 | 3, 4 | 648 | ||
| XM_006250361.2 | XP_006250423.1 | 3, 4 | 690 | |||
| XM_008769817.1 | XP_008768039.1 | 3, 4 | 692 | |||
| GenBank mRNA | BC085870.1 | - | 3, 4 | 648 | 7191198 | |

Tfb2m
Genomic Map