siRNA Details
| Gene Symbol: Fam213a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008297.1 | NP_001008298.1 | 4 | 706 | ||
| XM_006252573.1 | XP_006252635.1 | 4 | 654 | |||
| GenBank mRNA | BC086323.1 | - | 4 | 706 | 7304524 | |
| FQ213064.1 | - | 540 | ||||
| FQ220194.1 | - | 4 | 533 | |||

Fam213a
Genomic Map