siRNA Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008302.1 | NP_001008303.1 | 2 | 168 | ||
| GenBank mRNA | BC085346.1 | - | 2 | 168 | 7304630 | |
| FQ225074.1 | - | 2 | 189 | |||
| FQ227930.1 | - | 2 | 146 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sft2d1| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008302.1 | NP_001008303.1 | 2 | 168 | ||
| GenBank mRNA | BC085346.1 | - | 2 | 168 | 7304630 | |
| FQ225074.1 | - | 2 | 189 | |||
| FQ227930.1 | - | 2 | 146 | |||