siRNA Details
| Gene Symbol: Snrpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008303.1 | NP_001008304.1 | 6 | 706 | ||
| XM_006228564.2 | XP_006228626.1 | 6 | 837 | |||
| GenBank mRNA | BC086331.1 | - | 6 | 706 | 7300752 | |
| FQ227089.1 | - | 5 | 600 | |||
| FQ231860.1 | - | 4 | 410 | |||
| FQ233559.1 | - | 5 | 600 | |||

Snrpa
Genomic Map