siRNA Details
| Gene Symbol: Zufsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008308.1 | NP_001008309.1 | 9 | 1602 | ||
| GenBank mRNA | BC085885.1 | - | 9 | 1602 | 7192192 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zufsp| Gene Symbol: Zufsp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008308.1 | NP_001008309.1 | 9 | 1602 | ||
| GenBank mRNA | BC085885.1 | - | 9 | 1602 | 7192192 | |