siRNA Details
Gene Symbol: Ermn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008311.3 | NP_001008312.1 | 3 | 877 | ||
GenBank mRNA | BC086596.1 | - | 3 | 877 | 7311836 | |
DQ119821.1 | - | 805 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ermn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008311.3 | NP_001008312.1 | 3 | 877 | ||
GenBank mRNA | BC086596.1 | - | 3 | 877 | 7311836 | |
DQ119821.1 | - | 805 |