siRNA Details
| Gene Symbol: Serinc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008312.1 | NP_001008313.1 | 8 | 1041 | ||
| GenBank mRNA | BC085853.1 | - | 8 | 1041 | 7190015 | |
| EU220431.1 | - | 8 | 987 | |||
| FQ222836.1 | - | 2 | 124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serinc3| Gene Symbol: Serinc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008312.1 | NP_001008313.1 | 8 | 1041 | ||
| GenBank mRNA | BC085853.1 | - | 8 | 1041 | 7190015 | |
| EU220431.1 | - | 8 | 987 | |||
| FQ222836.1 | - | 2 | 124 | |||