siRNA Details
| Gene Symbol: Ltbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008315.1 | NP_001008316.1 | 6, 7 | 855 | ||
| XM_006237351.1 | XP_006237413.1 | 6, 7 | 902 | |||
| GenBank mRNA | BC085880.1 | - | 6, 7 | 855 | 7191913 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltbr| Gene Symbol: Ltbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008315.1 | NP_001008316.1 | 6, 7 | 855 | ||
| XM_006237351.1 | XP_006237413.1 | 6, 7 | 902 | |||
| GenBank mRNA | BC085880.1 | - | 6, 7 | 855 | 7191913 | |