siRNA Details
| Gene Symbol: Ndufaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008318.1 | NP_001008319.1 | 8, 9 | 940 | ||
| XM_006239627.2 | XP_006239689.1 | 8, 9 | 1047 | |||
| GenBank mRNA | BC083810.1 | - | 8, 9 | 940 | 7110144 | |

Ndufaf7
Genomic Map