siRNA Details
| Gene Symbol: Gadd45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008321.1 | NP_001008322.1 | 4 | 724 | ||
| XM_006240888.2 | XP_006240950.1 | 3 | 975 | |||
| GenBank mRNA | BC085337.1 | - | 4 | 724 | 7302492 | |
| FQ235047.1 | - | 3 | 975 | |||

Gadd45b
Genomic Map