siRNA Details
| Gene Symbol: Pmm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008323.1 | NP_001008324.1 | 4 | 347 | ||
| GenBank mRNA | BC086346.1 | - | 4 | 347 | 7302298 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pmm1| Gene Symbol: Pmm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008323.1 | NP_001008324.1 | 4 | 347 | ||
| GenBank mRNA | BC086346.1 | - | 4 | 347 | 7302298 | |