siRNA Details
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008324.1 | NP_001008325.1 | 12, 13 | 1583 | ||
GenBank mRNA | BC085933.1 | - | 12, 13 | 1583 | 7112408 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008324.1 | NP_001008325.1 | 12, 13 | 1583 | ||
GenBank mRNA | BC085933.1 | - | 12, 13 | 1583 | 7112408 |