siRNA Details
Gene Symbol: Tmem218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008325.1 | NP_001008326.1 | 4 | 756 | ||
XM_006242802.2 | XP_006242864.1 | 4 | 871 | |||
XM_008766050.1 | XP_008764272.1 | 4 | 907 | |||
GenBank mRNA | BC085342.1 | - | 4 | 756 | 7303171 | |
FQ214300.1 | - | 4 | 776 |
