siRNA Details
| Gene Symbol: ARHGEF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014784.3 | NP_055599.1 | 6, 7 | 1558 | ||
| NM_198236.2 | NP_937879.1 | 6, 7 | 1558 | |||
| XM_005245629.4 | XP_005245686.1 | 6, 7 | 1959 | |||
| XM_006711659.2 | XP_006711722.1 | 8, 9 | 2007 | |||
| XM_006711660.3 | XP_006711723.1 | 8, 9 | 2007 | |||
| XM_006711661.2 | XP_006711724.1 | 8, 9 | 2007 | |||
| XM_006711662.2 | XP_006711725.1 | 8, 9 | 2007 | |||
| XM_006711663.3 | XP_006711726.1 | 6, 7 | 994 | |||
| XM_006711665.3 | XP_006711728.1 | 8, 9 | 2007 | |||
| XM_011510185.2 | XP_011508487.1 | 7, 8 | 1992 | |||
| XM_011510186.2 | XP_011508488.1 | 8, 9 | 2007 | |||
| XM_011510187.2 | XP_011508489.1 | 7, 8 | 718 | |||
| XM_017002920.1 | XP_016858409.1 | 7, 8 | 1974 | |||
| XM_017002921.1 | XP_016858410.1 | 6, 7 | 1959 | |||
| XM_017002922.1 | XP_016858411.1 | 8, 9 | 2007 | |||
| XM_017002923.1 | XP_016858412.1 | 6, 7 | 1959 | |||
| XM_017002924.1 | XP_016858413.1 | 7, 8 | 1992 | |||
| GenBank mRNA | AB002378.1 | - | 6, 7 | 1263 | ||
| BC057394.1 | - | 6, 7 | 1582 | 6148684 | ||

ARHGEF11
Genomic Map