siRNA Details
| Gene Symbol: Amn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008333.2 | NP_001008334.2 | 4 | 405 | ||
| XM_006237700.1 | XP_006237762.1 | 4 | 891 | |||
| GenBank mRNA | BC086357.1 | - | 4 | 375 | 7303722 | |
| BG672304.1 | - | 4 | 405 | |||

Amn1
Genomic Map