siRNA Details
| Gene Symbol: Amn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008333.2 | NP_001008334.2 | 5 | 628 | ||
| XM_006237700.1 | XP_006237762.1 | 5 | 1114 | |||
| GenBank mRNA | BC086357.1 | - | 5 | 598 | 7303722 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amn1| Gene Symbol: Amn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008333.2 | NP_001008334.2 | 5 | 628 | ||
| XM_006237700.1 | XP_006237762.1 | 5 | 1114 | |||
| GenBank mRNA | BC086357.1 | - | 5 | 598 | 7303722 | |