siRNA Details
| Gene Symbol: Eif4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008335.1 | NP_001008336.1 | 11 | 1606 | ||
| XM_006248534.2 | XP_006248596.1 | 11 | 1622 | |||
| GenBank mRNA | BC085859.1 | - | 11 | 1606 | 7190333 | |
| FQ215479.1 | - | 4 | 759 | |||

Eif4a2
Genomic Map